siRNA Details
| Gene Symbol: Ddah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165936.1 | NP_001159408.1 | 2 | 339 | ||
| NM_212532.2 | NP_997697.1 | 2 | 386 | |||
| XM_008772714.1 | XP_008770936.1 | 3 | 485 | |||
| XM_008772715.1 | XP_008770937.1 | 2 | 648 | |||
| GenBank mRNA | BC086443.1 | - | 2 | 287 | 7303579 | |
| BC097930.1 | - | 2 | 376 | 7388716 | ||
| CO385374.1 | - | 2 | 386 | |||
| EV770973.1 | - | 2 | 339 | |||

Ddah2
Genomic Map