siRNA Details
| Gene Symbol: Htt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024357.3 | NP_077333.2 | 41 | 5536 | ||
| XM_006251230.2 | XP_006251292.1 | 40 | 5474 | |||
| XM_006251232.2 | XP_006251294.1 | 41 | 5522 | |||
| XM_008770274.1 | XP_008768496.1 | 39 | 4988 | |||
| GenBank mRNA | U01022.1 | - | 1 | 24 | ||
| U18650.1 | - | 41 | 5380 | |||

Htt
Genomic Map