siRNA Details
| Gene Symbol: Psmb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105727.2 | NP_001099197.2 | 3 | 623 | ||
| GenBank mRNA | AI602737.1 | - | 1 | 240 | ||
| D45247.1 | - | 3 | 621 | |||
| FQ214283.1 | - | 2 | 157 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmb5| Gene Symbol: Psmb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105727.2 | NP_001099197.2 | 3 | 623 | ||
| GenBank mRNA | AI602737.1 | - | 1 | 240 | ||
| D45247.1 | - | 3 | 621 | |||
| FQ214283.1 | - | 2 | 157 | |||