siRNA Details
| Gene Symbol: Cuta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164706.1 | NP_001158178.1 | 2 | 133 | ||
| NM_001164707.1 | NP_001158179.1 | 1 | 186 | |||
| NM_212494.2 | NP_997659.2 | 2 | 186 | |||
| XM_006256120.2 | XP_006256182.1 | 3 | 217 | |||
| GenBank mRNA | BP501235.1 | - | 1 | 200 | ||
| CB586341.1 | - | 2 | 133 | |||
| DV717217.1 | - | 1 | 158 | |||
| FQ220708.1 | - | 2 | 420 | |||

Cuta
Genomic Map