siRNA Details
| Gene Symbol: RGD1564450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106377.1 | NP_001099847.1 | 5 | 787 | ||
| XM_006256171.2 | XP_006256233.1 | 5 | 605 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564450| Gene Symbol: RGD1564450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106377.1 | NP_001099847.1 | 5 | 787 | ||
| XM_006256171.2 | XP_006256233.1 | 5 | 605 | |||