siRNA Details
| Gene Symbol: Nudt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024243.1 | NP_001019414.1 | 5 | 695 | ||
| GenBank mRNA | BC093618.1 | - | 5 | 695 | 7311727 | |
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Nudt3
Nudt3| Gene Symbol: Nudt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024243.1 | NP_001019414.1 | 5 | 695 | ||
| GenBank mRNA | BC093618.1 | - | 5 | 695 | 7311727 | |
 Back To Top
 Back To Top