siRNA Details
Gene Symbol: Fam207a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008307.1 | NP_001008308.1 | 2 | 261 | ||
GenBank mRNA | BC083697.1 | - | 2 | 261 | 7189806 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam207a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008307.1 | NP_001008308.1 | 2 | 261 | ||
GenBank mRNA | BC083697.1 | - | 2 | 261 | 7189806 |