siRNA Details
| Gene Symbol: Fam207a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008307.1 | NP_001008308.1 | 5 | 556 | ||
| GenBank mRNA | BC083697.1 | - | 5 | 556 | 7189806 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam207a| Gene Symbol: Fam207a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008307.1 | NP_001008308.1 | 5 | 556 | ||
| GenBank mRNA | BC083697.1 | - | 5 | 556 | 7189806 | |