siRNA Details
| Gene Symbol: Pcbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011945.1 | NP_001011945.1 | 13, 14 | 1217 | ||
| XM_006256283.2 | XP_006256345.1 | 14, 15 | 1396 | |||
| XM_006256285.2 | XP_006256347.1 | 11, 12 | 980 | |||
| XM_006256286.2 | XP_006256348.1 | 14, 15 | 1393 | |||
| XM_006256287.2 | XP_006256349.1 | 13, 14 | 1337 | |||
| XM_006256288.2 | XP_006256350.1 | 14, 15 | 1325 | |||
| XM_006256289.2 | XP_006256351.1 | 14, 15 | 1322 | |||
| XM_006256290.2 | XP_006256352.1 | 12, 13 | 2395 | |||
| XM_006256291.2 | XP_006256353.1 | 13, 14 | 1266 | |||
| XM_008772874.1 | XP_008771096.1 | 12, 13 | 2416 | |||
| XM_008772875.1 | XP_008771097.1 | 12, 13 | 1100 | |||
| GenBank mRNA | BC079196.1 | - | 13, 14 | 1217 | 7131426 | |

Pcbp3
Genomic Map