siRNA Details
Gene Symbol: Col6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_215375.7 | XP_215375.5 | 35 | 2854 | ||
GenBank mRNA | FQ224671.1 | - | 2 | 265 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Col6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_215375.7 | XP_215375.5 | 35 | 2854 | ||
GenBank mRNA | FQ224671.1 | - | 2 | 265 |