siRNA Details
| Gene Symbol: Pak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019210.1 | NP_062083.1 | 13, 14 | 1477 | ||
| XM_006257333.2 | XP_006257395.1 | 15, 16 | 2750 | |||
| XM_006257336.2 | XP_006257398.1 | 14, 15 | 2639 | |||
| XM_006257338.2 | XP_006257400.1 | 14, 15 | 3042 | |||
| XM_006257339.2 | XP_006257401.1 | 14, 15 | 2704 | |||
| XM_008773404.1 | XP_008771626.1 | 16, 17 | 2815 | |||
| GenBank mRNA | U33314.1 | - | 13, 14 | 1477 | ||

Pak3
Genomic Map