siRNA Details
| Gene Symbol: Ssr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017199.1 | NP_058895.1 | 2 | 264 | ||
| XM_006229571.2 | XP_006229633.1 | 2 | 320 | |||
| GenBank mRNA | BC058482.1 | - | 2 | 177 | 6921620 | |
| Z19087.1 | - | 2 | 264 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ssr4| Gene Symbol: Ssr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017199.1 | NP_058895.1 | 2 | 264 | ||
| XM_006229571.2 | XP_006229633.1 | 2 | 320 | |||
| GenBank mRNA | BC058482.1 | - | 2 | 177 | 6921620 | |
| Z19087.1 | - | 2 | 264 | |||