siRNA Details
| Gene Symbol: Rfx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106388.1 | NP_001099858.1 | 14 | 1927 | ||
| XM_006256444.2 | XP_006256506.1 | 17 | 2229 | |||
| XM_006256445.2 | XP_006256507.1 | 10 | 1480 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rfx6| Gene Symbol: Rfx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106388.1 | NP_001099858.1 | 14 | 1927 | ||
| XM_006256444.2 | XP_006256506.1 | 17 | 2229 | |||
| XM_006256445.2 | XP_006256507.1 | 10 | 1480 | |||