siRNA Details
| Gene Symbol: Rfx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106388.1 | NP_001099858.1 | 9 | 919 | ||
| XM_006256444.2 | XP_006256506.1 | 12 | 1221 | |||
| XM_006256445.2 | XP_006256507.1 | 5 | 472 | |||
| Gene Symbol: LOC100911315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772970.1 | XP_008771192.1 | 1 | 154 | ||

Rfx6
Genomic Map