siRNA Details
| Gene Symbol: Fam26e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024977.1 | NP_001020148.1 | 1 | 253 | ||
| GenBank mRNA | BC089225.1 | - | 1 | 253 | 7309006 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam26e| Gene Symbol: Fam26e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024977.1 | NP_001020148.1 | 1 | 253 | ||
| GenBank mRNA | BC089225.1 | - | 1 | 253 | 7309006 | |