siRNA Details
| Gene Symbol: Ahcy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017201.1 | NP_058897.1 | 4, 5 | 467 | ||
| GenBank mRNA | BC061841.1 | - | 4, 5 | 494 | 5599994 | |
| M15185.1 | - | 4, 5 | 467 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ahcy| Gene Symbol: Ahcy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017201.1 | NP_058897.1 | 4, 5 | 467 | ||
| GenBank mRNA | BC061841.1 | - | 4, 5 | 494 | 5599994 | |
| M15185.1 | - | 4, 5 | 467 | |||