siRNA Details
| Gene Symbol: Ghrh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031577.1 | NP_113765.1 | 4 | 411 | ||
| XM_008762304.1 | XP_008760526.1 | 4 | 313 | |||
| GenBank mRNA | M73486.1 | - | 4 | 411 | ||
| U10156.1 | - | 4 | 313 | |||
| U41183.1 | - | 3 | 234 | |||
| Z34092.1 | - | 4 | 313 | |||

Ghrh
Genomic Map