siRNA Details
Gene Symbol: Rab7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023950.3 | NP_076440.1 | 3 | 277 | ||
XM_006236849.2 | XP_006236911.1 | 3 | 228 | |||
GenBank mRNA | AB158427.1 | - | 2 | 109 | ||
AB158428.1 | - | 2 | 109 | |||
AF286535.1 | - | 3 | 273 | |||
BC072470.1 | - | 3 | 277 | 7098657 | ||
X12535.1 | - | 3 | 150 |
