siRNA Details
| Gene Symbol: Mical1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106397.1 | NP_001099867.1 | 17 | 2479 | ||
| XM_008772984.1 | XP_008771206.1 | 17 | 4186 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mical1| Gene Symbol: Mical1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106397.1 | NP_001099867.1 | 17 | 2479 | ||
| XM_008772984.1 | XP_008771206.1 | 17 | 4186 | |||