siRNA Details
| Gene Symbol: Epas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023090.1 | NP_075578.1 | 16 | 2640 | ||
| GenBank mRNA | AJ277828.1 | - | 16 | 2640 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epas1| Gene Symbol: Epas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023090.1 | NP_075578.1 | 16 | 2640 | ||
| GenBank mRNA | AJ277828.1 | - | 16 | 2640 | ||