siRNA Details
| Gene Symbol: Gdf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019216.2 | NP_062089.1 | 2 | 571 | ||
| Gene Symbol: Lrrc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_573882.5 | XP_573882.1 | 4 | 3098 | ||
| GenBank mRNA | FQ228377.1 | - | 4 | 3079 | ||

Gdf15
Genomic Map