siRNA Details
| Gene Symbol: Map1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019217.1 | NP_062090.1 | 1 | 262 | ||
| GenBank mRNA | BC158858.1 | - | 1 | 175 | ||
| CB610210.1 | - | 1 | 262 | |||
| U52950.1 | - | 1 | 158 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map1b| Gene Symbol: Map1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019217.1 | NP_062090.1 | 1 | 262 | ||
| GenBank mRNA | BC158858.1 | - | 1 | 175 | ||
| CB610210.1 | - | 1 | 262 | |||
| U52950.1 | - | 1 | 158 | |||