siRNA Details
| Gene Symbol: RGD1560883 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231723.2 | XP_006231785.1 | 3 | 307 | ||
| XM_008760618.1 | XP_008758840.1 | 3 | 307 | |||
| XM_008760619.1 | XP_008758841.1 | 3 | 320 | |||
| XM_008760620.1 | XP_008758842.1 | 3 | 305 | |||
| XM_008760621.1 | XP_008758843.1 | 3 | 305 | |||
| XM_008760622.1 | XP_008758844.1 | 3 | 305 | |||

RGD1560883
Genomic Map