siRNA Details
| Gene Symbol: Otp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100523.1 | NP_001093993.1 | 2 | 496 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Otp| Gene Symbol: Otp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100523.1 | NP_001093993.1 | 2 | 496 | ||