siRNA Details
| Gene Symbol: Ankra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207595.1 | NP_997478.1 | 3, 4 | 652 | ||
| XM_006257186.2 | XP_006257248.1 | 2, 3 | 458 | |||
| GenBank mRNA | AY566568.1 | - | 3, 4 | 652 | ||
| BC081975.1 | - | 3, 4 | 841 | 7126874 | ||
| Gene Symbol: LOC100910438 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760690.1 | XP_008758912.1 | 3, 4 | 904 | ||
| XM_008760691.1 | XP_008758913.1 | 2, 3 | 458 | |||

Ankra2
Genomic Map