siRNA Details
| Gene Symbol: Dimt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106408.2 | NP_001099878.2 | 11 | 1042 | ||
| XM_006231902.2 | XP_006231964.1 | 11 | 1070 | |||
| XM_006231903.2 | XP_006231965.1 | 12 | 1253 | |||
| XM_006231905.2 | XP_006231967.1 | 10 | 956 | |||
| GenBank mRNA | BC158729.1 | - | 11 | 1042 | 9091721 | |

Dimt1
Genomic Map