siRNA Details
| Gene Symbol: Dimt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106408.2 | NP_001099878.2 | 4, 5 | 475 | ||
| XM_006231902.2 | XP_006231964.1 | 4, 5 | 503 | |||
| XM_006231903.2 | XP_006231965.1 | 5, 6 | 686 | |||
| XM_006231905.2 | XP_006231967.1 | 3, 4 | 389 | |||
| GenBank mRNA | BC158729.1 | - | 4, 5 | 475 | 9091721 | |

Dimt1
Genomic Map