siRNA Details
| Gene Symbol: Gpx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106411.1 | NP_001099881.1 | 3 | 560 | ||
| GenBank mRNA | FQ214900.1 | - | 3 | 534 | ||
| FQ220116.1 | - | 3 | 531 | |||
| FQ220265.1 | - | 3 | 540 | |||
| FQ220543.1 | - | 3 | 540 | |||
| FQ229597.1 | - | 3 | 534 | |||
| FQ229607.1 | - | 3 | 530 | |||
| FQ229635.1 | - | 3 | 534 | |||
| FQ229721.1 | - | 3 | 534 | |||
| FQ230033.1 | - | 3 | 534 | |||

Gpx8
Genomic Map