siRNA Details
| Gene Symbol: Mocs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007633.2 | NP_001007634.1 | 5, 6 | 674 | ||
| NM_001162413.1 | NP_001155885.1 | 6, 7 | 796 | |||
| XM_006231985.2 | XP_006232047.1 | 6, 7 | 742 | |||
| XM_006231986.2 | XP_006232048.1 | 5, 6 | 639 | |||
| GenBank mRNA | BC079181.1 | - | 6, 7 | 762 | 7130858 | |
| FM123325.1 | - | 5, 6 | 627 | |||
| FQ220090.1 | - | 6, 7 | 691 | |||
| FQ220210.1 | - | 5, 6 | 587 | |||
| FQ230755.1 | - | 5, 6 | 597 | |||
| FQ235152.1 | - | 6, 7 | 725 | |||

Mocs2
Genomic Map