siRNA Details
| Gene Symbol: Mrps30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106412.1 | NP_001099882.1 | 4, 5 | 1017 | ||
| XM_006231957.2 | XP_006232019.1 | 5, 6 | 1118 | |||
| GenBank mRNA | FQ223189.1 | - | 4, 5 | 1028 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrps30| Gene Symbol: Mrps30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106412.1 | NP_001099882.1 | 4, 5 | 1017 | ||
| XM_006231957.2 | XP_006232019.1 | 5, 6 | 1118 | |||
| GenBank mRNA | FQ223189.1 | - | 4, 5 | 1028 | ||