siRNA Details
| Gene Symbol: Odf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145005.1 | NP_001138477.1 | 12 | 1300 | ||
| NM_017213.2 | NP_058909.2 | 12 | 1426 | |||
| GenBank mRNA | AF053971.1 | - | 11 | 1527 | ||
| AF162756.2 | - | 1367 | ||||
| BC078857.1 | - | 12 | 1426 | 7112637 | ||
| U62821.1 | - | 10 | 1258 | |||
| X95272.1 | - | 12 | 1267 | |||

Odf2
Genomic Map