siRNA Details
| Gene Symbol: Krt18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053976.1 | NP_446428.1 | 6 | 1119 | ||
| GenBank mRNA | BC091275.1 | - | 6 | 1119 | 7383311 | |
| Gene Symbol: LOC683212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760823.1 | XP_008759045.1 | 2 | 1079 | ||

Krt18
Genomic Map