siRNA Details
Gene Symbol: Smc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031583.2 | NP_113771.2 | 2 | 181 | ||
GenBank mRNA | BE109731.1 | - | 2 | 209 | ||
FQ225069.1 | - | 1 | 18 | |||
FQ231827.1 | - | 1 | 18 | |||
FQ231919.1 | - | 2 | 193 | |||
FQ233317.1 | - | 2 | 193 | |||
FQ234437.1 | - | 2 | 193 | |||
U82626.1 | - | 2 | 148 |
