siRNA Details
| Gene Symbol: Smc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031583.2 | NP_113771.2 | 20 | 2317 | ||
| GenBank mRNA | BP486487.1 | - | 1 | 32 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smc3| Gene Symbol: Smc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031583.2 | NP_113771.2 | 20 | 2317 | ||
| GenBank mRNA | BP486487.1 | - | 1 | 32 | ||