siRNA Details
| Gene Symbol: Ralyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024978.1 | NP_001020149.1 | 5, 6 | 521 | ||
| XM_006232126.2 | XP_006232188.1 | 6, 7 | 1343 | |||
| GenBank mRNA | BC092614.1 | - | 5, 6 | 521 | 7311942 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ralyl| Gene Symbol: Ralyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024978.1 | NP_001020149.1 | 5, 6 | 521 | ||
| XM_006232126.2 | XP_006232188.1 | 6, 7 | 1343 | |||
| GenBank mRNA | BC092614.1 | - | 5, 6 | 521 | 7311942 | |