siRNA Details
| Gene Symbol: Itsn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136096.1 | NP_001129568.1 | 843 | |||
| NM_019227.1 | NP_062100.1 | 8, 9 | 822 | |||
| GenBank mRNA | AF127798.1 | - | 8, 9 | 822 | ||
| AF132672.1 | - | 8, 9 | 822 | |||

Itsn1
Genomic Map