siRNA Details
Gene Symbol: Mrpl47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037183.1 | NP_001032260.1 | 6 | 566 | ||
XM_008760912.1 | XP_008759134.1 | 5 | 403 | |||
GenBank mRNA | BC089884.1 | - | 6 | 567 | ||
BC105820.1 | - | 6 | 566 | 7369885 | ||
FQ222657.1 | - | 11 |
