siRNA Details
| Gene Symbol: Acad9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181768.2 | NP_861433.2 | 17 | 1764 | ||
| XM_006232269.1 | XP_006232331.1 | 17 | 1844 | |||
| GenBank mRNA | AY292988.1 | - | 4 | 361 | ||
| BC162016.1 | - | 17 | 1764 | 9092768 | ||

Acad9
Genomic Map