siRNA Details
| Gene Symbol: RGD1307100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037184.1 | NP_001032261.1 | 3 | 748 | ||
| XM_006232275.2 | XP_006232337.1 | 87 | 15208 | |||
| XM_008760914.1 | XP_008759136.1 | 87 | 15208 | |||
| XM_008760915.1 | XP_008759137.1 | 87 | 15205 | |||
| XM_008760916.1 | XP_008759138.1 | 86 | 15157 | |||
| XM_008760917.1 | XP_008759139.1 | 86 | 15145 | |||
| XM_008760918.1 | XP_008759140.1 | 86 | 15070 | |||
| XM_008760919.1 | XP_008759141.1 | 86 | 15006 | |||
| GenBank mRNA | BC100102.1 | - | 3 | 748 | 7377390 | |

RGD1307100
Genomic Map