siRNA Details
Gene Symbol: Adad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006977.2 | NP_001006978.2 | 3, 4 | 507 | ||
XM_006232278.2 | XP_006232340.1 | 3, 4 | 691 | |||
XM_006232279.2 | XP_006232341.1 | 4, 5 | 543 | |||
GenBank mRNA | BC105906.1 | - | 3, 4 | 573 | 7453283 | |
CV126304.1 | - | 3, 4 | 538 |
