siRNA Details
| Gene Symbol: Spry1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106427.1 | NP_001099897.1 | 2 | 331 | ||
| GenBank mRNA | BC169059.1 | - | 3 | 630 | 7938818 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spry1| Gene Symbol: Spry1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106427.1 | NP_001099897.1 | 2 | 331 | ||
| GenBank mRNA | BC169059.1 | - | 3 | 630 | 7938818 | |