siRNA Details
Gene Symbol: Slc22a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031584.2 | NP_113772.1 | 1 | 106 | ||
GenBank mRNA | D83044.1 | - | 1 | 109 | ||
X98334.1 | - | 1 | 55 | |||
Y13154.1 | - | 1 | 56 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc22a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031584.2 | NP_113772.1 | 1 | 106 | ||
GenBank mRNA | D83044.1 | - | 1 | 109 | ||
X98334.1 | - | 1 | 55 | |||
Y13154.1 | - | 1 | 56 |