siRNA Details
| Gene Symbol: Exosc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106432.1 | NP_001099902.1 | 9 | 1099 | ||
| XM_006232332.2 | XP_006232394.1 | 10 | 717 | |||
| GenBank mRNA | FQ230358.1 | - | 9 | 664 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Exosc8| Gene Symbol: Exosc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106432.1 | NP_001099902.1 | 9 | 1099 | ||
| XM_006232332.2 | XP_006232394.1 | 10 | 717 | |||
| GenBank mRNA | FQ230358.1 | - | 9 | 664 | ||