siRNA Details
| Gene Symbol: Schip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100666.1 | NP_001094136.1 | 7 | 1181 | ||
| XM_008761079.1 | XP_008759301.1 | 4 | 4119 | |||
| XM_008761080.1 | XP_008759302.1 | 4 | 988 | |||
| XM_008761081.1 | XP_008759303.1 | 4 | 864 | |||
| XM_008761082.1 | XP_008759304.1 | 3 | 3136 | |||
| XM_008761083.1 | XP_008759305.1 | 3 | 435 | |||
| GenBank mRNA | BC092618.1 | - | 3 | 153 | ||
| BC169016.1 | - | 3 | 382 | |||

Schip1
Genomic Map