siRNA Details
| Gene Symbol: Lgals7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022582.2 | NP_072104.2 | 2 | 268 | ||
| XM_006228668.2 | XP_006228730.1 | 5 | 526 | |||
| XM_006228670.2 | XP_006228732.1 | 4 | 698 | |||
| GenBank mRNA | AF036941.1 | - | 2 | 277 | ||
| BI278774.1 | - | 2 | 257 | |||
| U67883.1 | - | 3 | 277 | |||

Lgals7
Genomic Map