siRNA Details
| Gene Symbol: Lgals7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022582.2 | NP_072104.2 | 3 | 323 | ||
| XM_006228668.2 | XP_006228730.1 | 6 | 581 | |||
| XM_006228670.2 | XP_006228732.1 | 5 | 753 | |||
| GenBank mRNA | AF036941.1 | - | 3 | 332 | ||
| BI278774.1 | - | 1 | 202 | |||
| U67883.1 | - | 4 | 332 | |||

Lgals7
Genomic Map