siRNA Details
| Gene Symbol: Krtcap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106444.1 | NP_001099914.1 | 2, 3 | 666 | ||
| GenBank mRNA | BC167108.1 | - | 2, 3 | 227 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtcap2| Gene Symbol: Krtcap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106444.1 | NP_001099914.1 | 2, 3 | 666 | ||
| GenBank mRNA | BC167108.1 | - | 2, 3 | 227 | ||