siRNA Details
| Gene Symbol: Mllt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013912.1 | NP_001013934.1 | 2 | 52 | ||
| GenBank mRNA | BC087583.1 | - | 2 | 52 | 7316660 | |
| BC094215.1 | - | 2 | 52 | 7318415 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mllt11| Gene Symbol: Mllt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013912.1 | NP_001013934.1 | 2 | 52 | ||
| GenBank mRNA | BC087583.1 | - | 2 | 52 | 7316660 | |
| BC094215.1 | - | 2 | 52 | 7318415 | ||