siRNA Details
| Gene Symbol: Hpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017233.1 | NP_058929.1 | 9 | 616 | ||
| GenBank mRNA | AF082834.1 | - | 9 | 616 | ||
| BC081819.1 | - | 9 | 614 | 7107086 | ||
| FQ209393.1 | - | 8 | 771 | |||
| FQ209489.1 | - | 8 | 576 | |||
| FQ210905.1 | - | 9 | 624 | |||
| FQ218378.1 | - | 9 | 624 | |||
| FQ218745.1 | - | 9 | 623 | |||
| FQ219696.1 | - | 9 | 624 | |||
| M18405.1 | - | 8 | 572 | |||

Hpd
Genomic Map