siRNA Details
| Gene Symbol: Ppm1j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005540.1 | NP_001005540.1 | 7, 8 | 1115 | ||
| XM_008761380.1 | XP_008759602.1 | 8, 9 | 1613 | |||
| GenBank mRNA | BC082053.1 | - | 7, 8 | 1115 | 7132147 | |

Ppm1j
Genomic Map